3-Bromobenzonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: m-Bromobenzonitrile

OR11089-100G OR11089-25G
APOSOR11089-100GEA 40.8 EUR
APOSOR11089-100G APOSOR11089-25G
3-Bromobenzonitrile 98%
3-Bromobenzonitrile
Formula: C₇H₄BrN
MW: 182,02 g/mol
Boiling Pt: 224…226 °C
Melting Pt: 37…41 °C
Density: 1,562 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
EINECS: 230-127-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR