Methyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate

Supplier: Apollo Scientific
Warning

OR110968-1G OR110968-5G OR110968-250MG
APOSOR110968-1GEA 27.7 EUR
APOSOR110968-1G APOSOR110968-5G APOSOR110968-250MG
Methyl-6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Formula: C₉H₇ClN₂O₂
MW: 210,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07021508
CAS Number: 900019-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR