N-Methylfufurylamine

Supplier: Apollo Scientific

Synonyms: N-Methylfurfurylamine

OR110977-1G OR110977-5G
APOSOR110977-1GEA 80 EUR
APOSOR110977-1G APOSOR110977-5G
N-Methylfufurylamine
N-Methylfufurylamine
Formula: C₆H₉NO
MW: 111,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020961
CAS Number: 4753-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR