4-Bromo-1-trityl-1H-Pyrazole

Supplier: Apollo Scientific

Synonyms: 4-Bromo-1-tritylpyrazole

OR111071-1G OR111071-5G
APOSOR111071-1GEA 64 EUR
APOSOR111071-1G APOSOR111071-5G
4-Bromo-1-trityl-1H-Pyrazole
4-Bromo-1-trityl-1H-Pyrazole
Formula: C₂₂H₁₇BrN₂
MW: 389,296 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907863
CAS Number: 95162-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR