2-(Chloromethyl)-5-methylbenzoxazole

Supplier: Apollo Scientific
Warning

OR111202-5G OR111202-1G
APOSOR111202-5GEA 227 EUR
APOSOR111202-5G APOSOR111202-1G
2-(Chloromethyl)-5-methylbenzoxazole
2-(Chloromethyl)-5-methylbenzoxazole
Formula: C₉H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD07366537
CAS Number: 41014-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR