9-Anthrealdehyde

Supplier: Apollo Scientific
Warning

Synonyms: 9-Anthracenecarbaldehyde, 9-Anthracenecarboxaldehyde, 9-Anthraldehyde

OR11122-100G OR11122-25G OR11122-500G
APOSOR11122-100GEA 63.8 EUR
APOSOR11122-100G APOSOR11122-25G APOSOR11122-500G
9-Anthrealdehyde
9-Anthrealdehyde
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 369
Melting Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00001254
CAS Number: 642-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR