2-(4-Methylpiperazin-1-yl)isonicotinonitrile
Supplier: Apollo Scientific
OR111296-5G
OR111296-1G
APOSOR111296-5GEA
171
EUR
APOSOR111296-5G
APOSOR111296-1G
2-(4-Methylpiperazin-1-yl)isonicotinonitrile
2-(4-Methylpiperazin-1-yl)isonicotinonitrile
Formula:
C₁₁H₁₄N₄ Storage Temperature: Ambient |
MDL Number:
MFCD09932504 CAS Number: 1016501-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...