6-Chloroquinoxaline
Supplier: Apollo Scientific
|
Danger
|
OR11146-1G
OR11146-25G
OR11146-5G
APOSOR11146-1GEA
21.9
EUR
APOSOR11146-1G
APOSOR11146-25G
APOSOR11146-5G
6-Chloroquinoxaline
6-Chloroquinoxaline
Formula:
C₈H₅ClN₂ MW: 164.5917 g/mol Boiling Pt: 117…119 °C Melting Pt: 61…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047594 CAS Number: 5448-43-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...