(Iodomethyl)cyclopropane 95%

Supplier: Apollo Scientific

OR111605-10G OR111605-1G OR111605-5G
APOSOR111605-10GEA 522 EUR
APOSOR111605-10G APOSOR111605-1G APOSOR111605-5G
(Iodomethyl)cyclopropane 95%
(Iodomethyl)cyclopropane
Formula: C₄H₇I
Boiling Pt: 53…54 °C
Storage Temperature: Ambient
MDL Number: MFCD06410734
CAS Number: 33574-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR