4-(Dimethylamino)benzoyl chloride
Supplier: Apollo Scientific
|
Danger
|
OR11167-25G
APOSOR11167-25GEA
150
EUR
APOSOR11167-25G
4-(Dimethylamino)benzoyl chloride
4-(Dimethylamino)benzoyl chloride
Formula:
C₉H₁₀ClNO MW: 183,64 g/mol Boiling Pt: 180 °C/15mmHg Melting Pt: 142…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236039 CAS Number: 4755-50-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...