2-Methoxybenzonitrile 97%

Supplier: Apollo Scientific
Warning

OR11202-100G OR11202-500G OR11202-25G
APOSOR11202-100GEA 80.1 EUR
APOSOR11202-100G APOSOR11202-500G APOSOR11202-25G
2-Methoxybenzonitrile 97%
2-Methoxybenzonitrile
Formula: C₈H₇NO
Boiling Pt: 256 °C
Density: 1,093 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001783
CAS Number: 6609-56-9
EINECS: 229-559-0
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR