4-Methyl-3-nitroanisole

Supplier: Apollo Scientific
Warning

Synonyms: 1-Methoxy-4-methyl-3-nitrobenzene, 4-Methoxy-2-nitrotoluene, Methyl 3-nitro-p-tolyl ether

OR11214-25G OR11214-5G
APOSOR11214-25GEA 50.9 EUR
APOSOR11214-25G APOSOR11214-5G
4-Methyl-3-nitroanisole
4-Methyl-3-nitroanisole
Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 266…267 °C
Melting Pt: 17 °C
Density: 1.207 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007173
CAS Number: 17484-36-5
EINECS: 241-500-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR