5-Nitroindan

Supplier: Apollo Scientific
Warning

Synonyms: 5-Nitroindane

OR11226-25G OR11226-5G
APOSOR11226-25GEA 441 EUR
APOSOR11226-25G APOSOR11226-5G
5-Nitroindan
5-Nitroindan
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 152
Melting Pt: 39
Density: 1.346
Storage Temperature: Ambient
CAS Number: 7436-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR