2-Chloro-4,6-dimethylpyrimidine

Supplier: Apollo Scientific
Warning

OR1125-100G
APOSOR1125-100GEA 163 EUR
APOSOR1125-100G
2-Chloro-4,6-dimethylpyrimidine
2-Chloro-4,6-dimethylpyrimidine
Formula: C₆H₇ClN₂
MW: 142,59 g/mol
Boiling Pt: 210…212 °C
Melting Pt: 34…38 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023199
CAS Number: 4472-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR