ω-Bromo-4-hydroxyacetophenone 97%

Supplier: Apollo Scientific
Danger

Synonyms: 2-Bromo-1-(4-hydroxyphenyl)ethanone, 2-Bromo-4'-hydroxyacetophenone, 2-Bromo-1-(4-hydroxyphenyl)ethan-1-one, α-Bromo-4-hydroxyacetophenone, 4-Hydroxyphenacyl bromide

OR11266-100G OR11266-25G
APOSOR11266-100GEA 69.8 EUR
APOSOR11266-100G APOSOR11266-25G
ω-Bromo-4-hydroxyacetophenone 97%
ω-Bromo-4-hydroxyacetophenone
Formula: C₈H₇BrO₂
MW: 215,05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00072424
CAS Number: 2491-38-5
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR