4-Phenyl-1,2,3-thiadiazol-5-amine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Amino-4-phenyl-1,2,3-thiadiazole

OR11285-1G OR11285-250MG
APOSOR11285-1GEA 141 EUR
APOSOR11285-1G APOSOR11285-250MG
4-Phenyl-1,2,3-thiadiazol-5-amine 97%
4-Phenyl-1,2,3-thiadiazol-5-amine
Formula: C₈H₇N₃S
MW: 177,23 g/mol
Melting Pt: 146…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00100232
CAS Number: 78301-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR