(2-Bromophenyl)acetic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(2-Bromophenyl)acetic acid
OR11296-100G
OR11296-25G
OR11296-500G
APOSOR11296-100GEA
90.2
EUR
APOSOR11296-100G
APOSOR11296-25G
APOSOR11296-500G
(2-Bromophenyl)acetic acid 98%
(2-Bromophenyl)acetic acid
Formula:
(C₆H₄Br)CH₂COOH MW: 215,05 g/mol Melting Pt: 104…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004314 CAS Number: 18698-97-0 EINECS: 242-509-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...