4-(2-Methoxyphenoxy)aniline
Supplier: Apollo Scientific
|
Warning
|
OR11344-25G
OR11344-5G
OR11344-100G
APOSOR11344-25GEA
87.3
EUR
APOSOR11344-25G
APOSOR11344-5G
APOSOR11344-100G
4-(2-Methoxyphenoxy)aniline
4-(2-Methoxyphenoxy)aniline
Formula:
C₁₃H₁₃NO₂ MW: 215,25 g/mol Melting Pt: 92,2…94.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD02663341 CAS Number: 13066-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...