Methyl-4-methoxypicolinate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 4-methoxypyridine-2-carboxylate

OR11373-1G OR11373-5G
APOSOR11373-1GEA 21.9 EUR
APOSOR11373-1G APOSOR11373-5G
Methyl-4-methoxypicolinate 97%
Methyl 4-methoxypicolinate
Formula: C₈H₉NO₃
MW: 167,16 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00956223
CAS Number: 29681-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR