6-Nitropicolinic acid 95%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Nitropyridine-2-carboxylic acid

OR11392-100MG OR11392-250MG
APOSOR11392-100MGEA 21.9 EUR
APOSOR11392-100MG APOSOR11392-250MG
6-Nitropicolinic acid 95%
6-Nitropicolinic acid
Formula: C₆H₄N₂O₄
MW: 168,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114205
CAS Number: 26893-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR