4-Nitrophenethylamine hydrochloride 98%

Supplier: Apollo Scientific

Synonyms: 2-(4-Nitrophenoxy)ethanamine hydrochloride, 4-(2-Aminoethoxy)-1-nitrobenzene hydrochloride

OR11412-1G OR11412-5G
APOSOR11412-1GEA 75.4 EUR
APOSOR11412-1G APOSOR11412-5G
4-Nitrophenethylamine hydrochloride 98%
4-Nitrophenethylamine hydrochloride
Formula: C₈H₁₁ClN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09475842
CAS Number: 98395-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR