2-Bromo-N-methylbenzenesulphonamide 98%

Supplier: Apollo Scientific
Warning

OR11471-1G OR11471-5G
APOSOR11471-1GEA 108 EUR
APOSOR11471-1G APOSOR11471-5G
2-Bromo-N-methylbenzenesulphonamide 98%
2-Bromo-N-methylbenzenesulphonamide
Formula: C₇H₈BrNO₂S
MW: 250,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03084756
CAS Number: 98192-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR