Isoscopoletin (6-hydroxy-4-methylcoumarin)

Supplier: Apollo Scientific
Warning

Synonyms: 6-Hydroxy-4-methyl-2H-chromen-2-one, 6-Hydroxy-4-methylcoumarin

OR1148-5G OR1148-1G
APOSOR1148-5GEA 39.4 EUR
APOSOR1148-5G APOSOR1148-1G
Isoscopoletin (6-hydroxy-4-methylcoumarin)
Isoscopoletin (6-Hydroxy-4-methylcoumarin)
Formula: C₁₀H₈O₃
MW: 176,17 g/mol
Melting Pt: 240…241 °C
Storage Temperature: Ambient
MDL Number: MFCD00016967
CAS Number: 2373-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR