7-Methoxy-4-methylcoumarin
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
7-Methoxy-4-methyl-2-benzopyrone, 4-Methylherniarin
OR1151-25G
OR1151-5G
OR1151-100G
APOSOR1151-25GEA
176
EUR
APOSOR1151-25G
APOSOR1151-5G
APOSOR1151-100G
7-Methoxy-4-methylcoumarin
7-Methoxy-4-methylcoumarin
Formula:
C₁₁H₁₀O₃ MW: 190,2 g/mol Melting Pt: 159…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009773 CAS Number: 2555-28-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...