Methyl-6-aminopicolinate 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Aminopicolinic acid methyll ester, Methyl 6-aminopicolinate, Methyl 6-Aminopicolinate, Methyl 6-Aminopyridine-2-carboxylate, 6-Aminopyridine-2-carboxylic acid methyll ester

OR11517-100G OR11517-25G OR11517-5G
APOSOR11517-100GEA 227 EUR
APOSOR11517-100G APOSOR11517-25G APOSOR11517-5G
Methyl-6-aminopicolinate 98%
Methyl 6-aminopicolinate
Formula: C₇H₈N₂O₂
MW: 152,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233712
CAS Number: 36052-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR