α-Naphthoflavone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Phenyl-4H-benzo[h]chromen-4-one, 2-Phenylbenzo[h]chromen-4-one, 7,8-Benzoflavone
OR1158-25G
APOSOR1158-25GEA
178
EUR
APOSOR1158-25G
α-Naphthoflavone
α-Naphthoflavone
Formula:
C₁₉H₁₂O₂ MW: 272,3 g/mol Melting Pt: 153…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004985 CAS Number: 604-59-1 EINECS: 210-071-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...