5-Bromo-4-isopropylpyrimidine 98%

Supplier: Apollo Scientific

OR11585-1G OR11585-5G
APOSOR11585-1GEA 208 EUR
APOSOR11585-1G APOSOR11585-5G
5-Bromo-4-isopropylpyrimidine 98%
5-Bromo-4-isopropylpyrimidine
Formula: C₇H₉BrN₂
MDL Number: MFCD09800930
CAS Number: 951884-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR