5-Bromo-4-phenylpyrimidine
Supplier: Apollo Scientific
|
Warning
|
OR11634-250MG
OR11634-1G
OR11634-5G
APOSOR11634-250MGEA
102
EUR
APOSOR11634-250MG
APOSOR11634-1G
APOSOR11634-5G
5-Bromo-4-phenylpyrimidine
5-Bromo-4-phenylpyrimidine
Formula:
C₁₀H₇BrN₂ MW: 235,0800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234599 CAS Number: 3543-46-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...