1-Methylindole

Supplier: Apollo Scientific
Warning

OR11669-100G OR11669-25G
APOSOR11669-100GEA 0 EUR
APOSOR11669-100G APOSOR11669-25G
1-Methylindole
1-Methylindole
Formula: C₉H₉N
Boiling Pt: 237…239 °C
Density: 1,051 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005800
CAS Number: 603-76-9
EINECS: 210-052-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR