5-Bromo-N-cyclopropylpicolinamide 98%

Supplier: Apollo Scientific
Warning

OR11915-1G OR11915-5G OR11915-25G
APOSOR11915-1GEA 144 EUR
APOSOR11915-1G APOSOR11915-5G APOSOR11915-25G
5-Bromo-N-cyclopropylpicolinamide 98%
5-Bromo-N-cyclopropylpicolinamide
Formula: C₉H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09801062
CAS Number: 638219-77-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR