N-Benzyl-4-bromobenzamide
Supplier: Apollo Scientific
|
Warning
|
OR11934-1G
OR11934-5G
APOSOR11934-1GEA
24.4
EUR
APOSOR11934-1G
APOSOR11934-5G
N-Benzyl-4-bromobenzamide
N-Benzyl-4-bromobenzamide
Formula:
C₁₄H₁₂BrNO Storage Temperature: Ambient |
MDL Number:
MFCD00443890 CAS Number: 80311-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...