4-Bromo-N-(prop-2-ynyl)benzamide 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Bromo-N-(prop-2-yn-1-yl)benzamide
OR11936-1G
OR11936-100MG
OR11936-5G
APOSOR11936-1GEA
241
EUR
APOSOR11936-1G
APOSOR11936-100MG
APOSOR11936-5G
4-Bromo-N-(prop-2-ynyl)benzamide 98%
4-Bromo-N-(prop-2-ynyl)benzamide
Formula:
C₁₀H₈BrNO MW: 238,0800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09800901 CAS Number: 82225-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...