1,4-Benzenediboronic acid bis(pinacol)ester
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1,4-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), 1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,4-Phenylenediboronic acid, pinacol ester
OR11953-1G
OR11953-5G
APOSOR11953-1GEA
51
EUR
APOSOR11953-1G
APOSOR11953-5G
1,4-Benzenediboronic acid bis(pinacol)ester
1,4-Benzenediboronic acid bis(pinacol)ester
Formula:
C₁₈H₂₈B₂O₄ MW: 330,04 g/mol Melting Pt: 241…245 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08276852 CAS Number: 99770-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...