trans-4-Pentylcyclohexanecarboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR11972-25G
OR11972-5G
APOSOR11972-25GEA
43.7
EUR
APOSOR11972-25G
APOSOR11972-5G
trans-4-Pentylcyclohexanecarboxylic acid
trans-4-Pentylcyclohexanecarboxylic acid
Formula:
C₁₁H₂₀O₂ MW: 198,30 g/mol Melting Pt: 51…53 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009964 CAS Number: 38289-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...