Methyl-5-bromo-4-chloro-2-hydroxybenzoate 98%

Supplier: Apollo Scientific
Warning

OR11997-1G OR11997-5G
APOSOR11997-1GEA 188 EUR
APOSOR11997-1G APOSOR11997-5G
Methyl-5-bromo-4-chloro-2-hydroxybenzoate 98%
Methyl 5-bromo-4-chloro-2-hydroxybenzoate
Formula: C₈H₆BrClO₃
Storage Temperature: Ambient
CAS Number: 55488-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR