5-Amino-2,2-dimethyl-pentan-1-ol 98%
Supplier: Apollo Scientific
|
Danger
|
OR12007-1G
APOSOR12007-1GEA
91.5
EUR
APOSOR12007-1G
5-Amino-2,2-dimethyl-pentan-1-ol 98%
5-Amino-2,2-dimethyl-pentan-1-ol
Formula:
C₇H₁₇NO MW: 131,22 g/mol Flash Pt: 108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043626 CAS Number: 13532-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...