3-Hydroxybenzaldehyde 98%
Supplier: Apollo Scientific
|
Warning
|
OR12009-100G
OR12009-1KG
OR12009-500G
APOSOR12009-100GEA
23.2
EUR
APOSOR12009-100G
APOSOR12009-1KG
APOSOR12009-500G
3-Hydroxybenzaldehyde 98%
3-Hydroxybenzaldehyde
Formula:
C₇H₆O₂ MW: 122,12 g/mol Boiling Pt: 191 °C/50mmHg Melting Pt: 100…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003368 CAS Number: 100-83-4 EINECS: 202-892-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...