1-Acetyl-5-indolinesulphonamide

Supplier: Apollo Scientific
Warning

OR12012-1G OR12012-5G
APOSOR12012-1GEA 125 EUR
APOSOR12012-1G APOSOR12012-5G
1-Acetyl-5-indolinesulphonamide
1-Acetyl-5-indolinesulphonamide
Formula: C₁₀H₁₂N₂O₃S
MW: 240.28 g/mol
Melting Pt: 225…227 °C
Storage Temperature: Ambient
MDL Number: MFCD01235910
CAS Number: 3264-38-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR