2-Amino-6,8-dibromo-3-quinolinecarbonitrile

Supplier: Apollo Scientific

OR12014-1G OR12014-5G
APOSOR12014-1GEA 219 EUR
APOSOR12014-1G APOSOR12014-5G
2-Amino-6,8-dibromo-3-quinolinecarbonitrile
2-Amino-6,8-dibromo-3-quinolinecarbonitrile
Formula: C₁₀H₅Br₂N₃
MW: 326,99 g/mol
Melting Pt: 313…315 °C
Storage Temperature: Ambient
MDL Number: MFCD08689714
CAS Number: 937601-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR