3'-{2-[4-(Benzyloxy)phenoxy]ethoxy}acetophenone
Supplier: Apollo Scientific
OR12027-10G
OR12027-1G
OR12027-5G
APOSOR12027-10GEA
902
EUR
APOSOR12027-10G
APOSOR12027-1G
APOSOR12027-5G
3'-{2-[4-(Benzyloxy)phenoxy]ethoxy}acetophenone
3'-{2-[4-(Benzyloxy)phenoxy]ethoxy}acetophenone
Formula:
C₂₃H₂₂O₄ Melting Pt: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689766 CAS Number: 937601-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...