8-Benzyl-1,2,8-triazaspiro[4.5]decan-3-one

Supplier: Apollo Scientific
Warning

OR12034-1G OR12034-5G
APOSOR12034-1GEA 118 EUR
APOSOR12034-1G APOSOR12034-5G
8-Benzyl-1,2,8-triazaspiro[4.5]decan-3-one
8-Benzyl-1,2,8-triazaspiro[4.5]decan-3-one
Formula: C₁₄H₁₉N₃O
Melting Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD08689731
CAS Number: 785782-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR