2-Amino-5-methylthiazole

Supplier: Apollo Scientific
Warning

Synonyms: 5-Methylthiazol-2-ylamine

OR1207-100G OR1207-25G
APOSOR1207-100GEA 21.9 EUR
APOSOR1207-100G APOSOR1207-25G
2-Amino-5-methylthiazole
2-Amino-5-methylthiazole
Formula: C₄H₆N₂S
MW: 114,17 g/mol
Melting Pt: 93…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00078317
CAS Number: 7305-71-7
EINECS: 423-800-5
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR