Ethyl-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[d]thiazole-4-carboxylate
OR12090-1G
OR12090-5G
APOSOR12090-1GEA
105
EUR
APOSOR12090-1G
APOSOR12090-5G
Ethyl-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate
Ethyl-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazole-4-carboxylate
Formula:
C₁₀H₁₄N₂O₂S MW: 226,3 g/mol Melting Pt: 173…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD08689777 CAS Number: 76263-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...