5-Bromo-2-methoxypyridine

Supplier: Apollo Scientific
Warning

OR1212-25G OR1212-100G
APOSOR1212-25GEA 98.9 EUR
APOSOR1212-25G APOSOR1212-100G
5-Bromo-2-methoxypyridine
5-Bromo-2-methoxypyridine
Formula: C₆H₆BrNO
MW: 188,02 g/mol
Boiling Pt: 80 °C/12mmHg
Density: 1,453 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01318952
CAS Number: 13472-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR