6-Methyl-2H-1,4-benzoxazine-2,3(4H)-dione 3-oxime
Supplier: Apollo Scientific
OR12122-10G
OR12122-1G
OR12122-5G
APOSOR12122-10GEA
597
EUR
APOSOR12122-10G
APOSOR12122-1G
APOSOR12122-5G
6-Methyl-2H-1,4-benzoxazine-2,3(4H)-dione 3-oxime
6-Methyl-2H-1,4-benzoxazine-2,3(4H)-dione 3-oxime
Formula:
C₉H₈N₂O₃ Melting Pt: 240…242 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD08689791 CAS Number: 937602-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...