5-(Chloromethyl)uracil

Supplier: Apollo Scientific
Warning

Synonyms: 5-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione

OR1213-100MG OR1213-250MG
APOSOR1213-100MGEA 19 EUR
APOSOR1213-100MG APOSOR1213-250MG
5-(Chloromethyl)uracil
5-(Chloromethyl)uracil
Formula: C₅H₅ClN₂O₂
MW: 160,56 g/mol
Melting Pt: >350 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00218445
CAS Number: 3590-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR