6-Methyl-5-nitro-1H-indazole

Supplier: Apollo Scientific
Warning

OR12145-1G OR12145-5G
APOSOR12145-1GEA 0 EUR
APOSOR12145-1G APOSOR12145-5G
6-Methyl-5-nitro-1H-indazole
6-Methyl-5-nitro-1H-indazole
Formula: C₈H₇N₃O₂
MW: 177.1601 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD03305368
CAS Number: 81115-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR