N-Methyl-3-nitro-2-pyridinamine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Methylamino)-3-nitropyridine
OR12147-1G
OR12147-5G
APOSOR12147-1GEA
72.7
EUR
APOSOR12147-1G
APOSOR12147-5G
N-Methyl-3-nitro-2-pyridinamine
N-Methyl-3-nitro-2-pyridinamine
Formula:
C₆H₇N₃O₂ MW: 153,14 g/mol Melting Pt: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00223335 CAS Number: 4093-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...