3-Methyl-1H-indazol-4-ol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
4-Hydroxy-3-methyl-1H-indazole
OR12205-1G
OR12205-250MG
APOSOR12205-1GEA
1080
EUR
APOSOR12205-1G
APOSOR12205-250MG
3-Methyl-1H-indazol-4-ol
3-Methyl-1H-indazol-4-ol
Formula:
C₈H₈N₂O MW: 148,17 g/mol Melting Pt: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027195 CAS Number: 149071-05-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...