1-(Piperidin-4-ylcarbonyl)piperidine 97%

Supplier: Apollo Scientific
Danger

OR12321-1G OR12321-250MG
APOSOR12321-1GEA 20.5 EUR
APOSOR12321-1G APOSOR12321-250MG
1-(Piperidin-4-ylcarbonyl)piperidine 97%
1-(Piperidin-4-ylcarbonyl)piperidine
Formula: C₁₁H₂₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD03372500
CAS Number: 63214-58-4
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR